D(-)-Quinic acid, 98+%
D(-)-Quinic acid, 98+%
D(-)-Quinic acid, 98+%
D(-)-Quinic acid, 98+%
Thermo Scientific Chemicals
D(-)-Quinic acid, 98+%
CAS: 77-95-2 | C7H12O6 | 192.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA160505000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD(-)-Quinic acid
CAS77-95-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Melting Point166.0°C to 168.0°C