N-Acetyl-L-cysteine, 98%
N-Acetyl-L-cysteine, 98%
N-Acetyl-L-cysteine, 98%
N-Acetyl-L-cysteine, 98%
Thermo Scientific Chemicals
N-Acetyl-L-cysteine, 98%
CAS: 616-91-1 | C5H9NO3S | 163.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
25 g
Packaging:
Glass bottle
Catalog number FSA160280500
Price (MYR)
456.00
BOT
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
456.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN-Acetyl-L-cysteine
CAS616-91-1
CAS Min %97.5
CAS Max %100.0
Melting Point109.0°C to 111.0°C