Agarose low EEO, for electrophoresis, (Mr <= 0.12)
Agarose low EEO, for electrophoresis, (Mr <= 0.12)
Agarose low EEO, for electrophoresis, (Mr <= 0.12)
Agarose low EEO, for electrophoresis, (Mr <= 0.12)
Thermo Scientific Chemicals
Agarose low EEO, for electrophoresis, (Mr <= 0.12)
CAS: 9012-36-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass Bottle
Catalog number FSA159821000
Price (MYR)
1,423.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,423.00
EA
Add to cart
Specifications
Chemical Name or MaterialAgarose low EEO
Physical FormFine Crystalline Powder
CAS9012-36-6
ColorWhite
EINECS Number232-731-8