N,N-Dimethylbenzylamine, 99%
N,N-Dimethylbenzylamine, 99%
N,N-Dimethylbenzylamine, 99%
N,N-Dimethylbenzylamine, 99%
Thermo Scientific Chemicals
N,N-Dimethylbenzylamine, 99%
CAS: 103-83-3 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA159755000
Price (MYR)
633.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
633.00
EA
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylbenzylamine
Name Note99%
CAS103-83-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger