4,4-Dimethyl-2-pentanone, 99%
4,4-Dimethyl-2-pentanone, 99%
4,4-Dimethyl-2-pentanone, 99%
4,4-Dimethyl-2-pentanone, 99%
Thermo Scientific Chemicals
4,4-Dimethyl-2-pentanone, 99%
CAS: 590-50-1 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number FSA159711000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material4, 4-Dimethyl-2-pentanone
CAS590-50-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0