2,3-Diaminonaphthalene, 97%
2,3-Diaminonaphthalene, 97%
2,3-Diaminonaphthalene, 97%
2,3-Diaminonaphthalene, 97%
Thermo Scientific Chemicals
2,3-Diaminonaphthalene, 97%
CAS: 771-97-1 | C10H10N2 | 158.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA159590010
Price (MYR)
1,321.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,321.00
BOT
Add to cart
Specifications
Chemical Name or Material2,3-Diaminonaphthalene
CAS771-97-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS91-59-8