Dimethyl malonate, 97%
Dimethyl malonate, 97%
Dimethyl malonate, 97%
Dimethyl malonate, 97%
Thermo Scientific Chemicals
Dimethyl malonate, 97%
CAS: 108-59-8 | C5H8O4 | 132.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA159560010
Price (MYR)
860.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
860.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDimethyl malonate
CAS108-59-8
Melting Point-62.0°C
ColorColorless
Density1.1560g/mL