1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals
1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals
1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals
1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA159450250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Iodo-2-nitrobenzene
Name Note97%
CAS609-73-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning