1,2-Dimethylimidazole, 96+%
1,2-Dimethylimidazole, 96+%
1,2-Dimethylimidazole, 96+%
1,2-Dimethylimidazole, 96+%
Thermo Scientific Chemicals
1,2-Dimethylimidazole, 96+%
CAS: 1739-84-0 | C5H8N2 | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA159445000
Price (MYR)
816.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
816.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 2-Dimethylimidazole
Name Note96+%
CAS1739-84-0
CAS Min %96+%
Health Hazard 1GHS Signal Word: Danger