Chalcone, 97%
Chalcone, 97%
Chalcone, 97%
Thermo Scientific Chemicals
Chalcone, 97%
CAS: 94-41-7 | C15H12O | 208.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA159431000
Price (MYR)
430.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
430.00
BOT
Add to cart
Specifications
Chemical Name or MaterialChalcone
Name Note97%
CAS94-41-7
Melting Point53.0°C to 58.0°C
ColorYellow