Catechol, 99+%
Catechol, 99+%
Catechol, 99+%
Catechol, 99+%
Thermo Scientific Chemicals
Catechol, 99+%
CAS: 120-80-9 | C6H6O2 | 110.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number FSA158985000
Price (MYR)
458.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
458.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCatechol
CAS120-80-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Suspected of causing genetic defects.
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.<b
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cau
View more
Suitable for battery materials development.
RUO – Research Use Only