sym-Diphenylcarbazide, 98%, ACS reagent
sym-Diphenylcarbazide, 98%, ACS reagent
sym-Diphenylcarbazide, 98%, ACS reagent
sym-Diphenylcarbazide, 98%, ACS reagent
Thermo Scientific Chemicals
sym-Diphenylcarbazide, 98%, ACS reagent
CAS: 140-22-7 | C13H14N4O | 242.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA158845000
Price (MYR)
4,184.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,184.00
EA
Add to cart
Specifications
Chemical Name or Materialsym-Diphenylcarbazide
CAS140-22-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g
View more
Indicator
RUO – Research Use Only