View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialVinylacetic acid
Name Notetech., 90%, unstabilized
CAS625-38-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: Causes severe skin burns and eye damage. Causes serious eye damage.