p-Xylene, 99%, pure
p-Xylene, 99%, pure
p-Xylene, 99%, pure
Thermo Scientific Chemicals
p-Xylene, 99%, pure
CAS: 106-42-3 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number FSA158775000
Price (MYR)
329.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
329.00
BOT
Add to cart
Specifications
Chemical Name or Materialp-Xylene
Melting Point13.0°C
CAS106-42-3
ColorColorless to Yellow
Health Hazard 1Danger