Succinic acid, 99%
Succinic acid, 99%
Succinic acid, 99%
Thermo Scientific Chemicals
Succinic acid, 99%
CAS: 110-15-6 | C4H6O4 | 118.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA158745000
Price (MYR)
434.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
434.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSuccinic acid
Name Note99%
CAS110-15-6
Melting Point185.0°C to 190.0°C
Boiling Point235.0°C