4-Phenylphenol, 97%
4-Phenylphenol, 97%
4-Phenylphenol, 97%
Thermo Scientific Chemicals
4-Phenylphenol, 97%
CAS: 92-69-3 | C12H10O | 170.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA158712500
Price (MYR)
437.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
437.00
EA
Add to cart
Specifications
Chemical Name or Material4-Phenylphenol
Name Note97%
CAS92-69-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning