Morpholine, 99+%, extra pure
Morpholine, 99+%, extra pure
Morpholine, 99+%, extra pure
Morpholine, 99+%, extra pure
Thermo Scientific Chemicals
Morpholine, 99+%, extra pure
Morpholine, 99+%, C4H9NO, CAS Number-110-91-8, 1-oxa-4-azacyclohexane, diethyleneimide oxide, diethylene imidoxide, diethylene oximide, diethylenimide oxide, drewamine, p-isoxazine, tetrahydro, tetrahydro-1,4-oxazine, tetrahydro-2h-1,4-oxazine, tetrahydro-p-oxazine, 1L, 99% min. (GC), 27, 5 | CAS: 110-91-8 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA158680010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMorpholine
CAS110-91-8
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0