trans-Cinnamic Acid, 98+%
trans-Cinnamic Acid, 98+%
trans-Cinnamic Acid, 98+%
trans-Cinnamic Acid, 98+%
Thermo Scientific Chemicals
trans-Cinnamic Acid, 98+%
CAS: 140-10-3 | C9H8O2 | 148.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number FSA158575000
Price (MYR)
584.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
584.00
BOT
Add to cart
Specifications
Chemical Name or Materialtrans-Cinnamic acid
Name Note98+%
CAS140-10-3
Melting Point131.0°C to 136.0°C
Boiling Point300.0°C