Aniline hydrochloride, 99%
Aniline hydrochloride, 99%
Aniline hydrochloride, 99%
Aniline hydrochloride, 99%
Thermo Scientific Chemicals
Aniline hydrochloride, 99%
CAS: 142-04-1 | C6H8ClN | 129.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA158411000
Price (MYR)
259.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
259.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAniline hydrochloride
Name Note99%
CAS142-04-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Suspected of causing cancer.
Suspected of causing genetic defects.
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction