2-(Methylamino)ethanol, 99%
2-(Methylamino)ethanol, 99%
2-(Methylamino)ethanol, 99%
2-(Methylamino)ethanol, 99%
Thermo Scientific Chemicals
2-(Methylamino)ethanol, 99%
CAS: 109-83-1 | C3H9NO | 75.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA158265000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-(Methylamino)ethanol
CAS109-83-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0