Bromoform, 96%, stabilized
Bromoform, 96%, stabilized
Bromoform, 96%, stabilized
Bromoform, 96%, stabilized
Thermo Scientific Chemicals
Bromoform, 96%, stabilized
CAS: 75-25-2 | CHBr3 | 252.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA158202500
Price (MYR)
1,010.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,010.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBromoform
CAS75-25-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
CAS64-17-5