2-Vinylpyridine, 97%, stabilized
2-Vinylpyridine, 97%, stabilized
2-Vinylpyridine, 97%, stabilized
2-Vinylpyridine, 97%, stabilized
Thermo Scientific Chemicals
2-Vinylpyridine, 97%, stabilized
CAS: 100-69-6 | C7H7N | 105.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
1 L
Packaging:
Glass bottle
Catalog number FSA158002500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Vinylpyridine
Name Note97%
CAS100-69-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger