Trifluoroacetic acid, sodium salt, 97%
Trifluoroacetic acid, sodium salt, 97%
Trifluoroacetic acid, sodium salt, 97%
Trifluoroacetic acid, sodium salt, 97%
Thermo Scientific Chemicals
Trifluoroacetic acid, sodium salt, 97%
CAS: 2923-18-4 | C2F3NaO2 | 136.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA157931000
Price (MYR)
977.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
977.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTrifluoroacetic acid, sodium salt
Name Note0.97
CAS2923-18-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement