1,2,4,5-Tetrachlorobenzene, 98%
1,2,4,5-Tetrachlorobenzene, 98%
1,2,4,5-Tetrachlorobenzene, 98%
1,2,4,5-Tetrachlorobenzene, 98%
Thermo Scientific Chemicals
1,2,4,5-Tetrachlorobenzene, 98%
CAS: 95-94-3 | C6H2Cl4 | 215.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA157780050
Price (MYR)
204.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
204.00
BOT
Add to cart
Specifications
Chemical Name or Material1,2,4,5-Tetrachlorobenzene
Melting Point139°C to 142°C
CAS95-94-3
CAS Min %97.5
ColorGray to Pink