1,10-Phenanthroline, 99+%
1,10-Phenanthroline, 99+%
1,10-Phenanthroline, 99+%
1,10-Phenanthroline, 99+%
Thermo Scientific Chemicals
1,10-Phenanthroline, 99+%
CAS: 66-71-7 | C12H8N2 | 180.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Catalog number FSA157535000
Price (MYR)
8,349.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
8,349.00
EA
Add to cart
Specifications
Chemical Name or Material1, 10-Phenanthroline
CAS66-71-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0