1-Nonanol, 97%
1-Nonanol, 97%
1-Nonanol, 97%
Thermo Scientific Chemicals
1-Nonanol, 97%
CAS: 143-08-8 | C9H20O | 144.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA157472500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Nonanol
CAS143-08-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0