N-Hydroxysuccinimide, 98+%
N-Hydroxysuccinimide, 98+%
N-Hydroxysuccinimide, 98+%
N-Hydroxysuccinimide, 98+%
Thermo Scientific Chemicals
N-Hydroxysuccinimide, 98+%
CAS: 6066-82-6 | C4H5NO3 | 115.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA157272500
Price (MYR)
1,356.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,356.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN-Hydroxysuccinimide
Name Note98+%
CAS6066-82-6
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
View more
Coupling reagent
RUO – Research Use Only