Methyltriphenylphosphonium Bromide, 98%
Methyltriphenylphosphonium Bromide, 98%
Methyltriphenylphosphonium Bromide, 98%
Methyltriphenylphosphonium Bromide, 98%
Thermo Scientific Chemicals
Methyltriphenylphosphonium Bromide, 98%
CAS: 1779-49-3 | C19H18BrP | 357.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
100 g
Packaging:
Plastic bottle
Catalog number FSA156955000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyltriphenylphosphonium bromide
CAS1779-49-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
View more
for C-C Bond Formation (Olefination)
RUO – Research Use Only