2-Mercaptopyrimidine, 98%
2-Mercaptopyrimidine, 98%
2-Mercaptopyrimidine, 98%
2-Mercaptopyrimidine, 98%
Thermo Scientific Chemicals
2-Mercaptopyrimidine, 98%
CAS: 1450-85-7 | C4H4N2S | 112.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA156740100
Price (MYR)
377.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
377.00
EA
Add to cart
Specifications
Chemical Name or Material2-Mercaptopyrimidine
Name Note98%
CAS1450-85-7
CAS Min %97.5
CAS Max %100.0