Malonaldehyde bis(diethyl acetal), 97%
Malonaldehyde bis(diethyl acetal), 97%
Malonaldehyde bis(diethyl acetal), 97%
Malonaldehyde bis(diethyl acetal), 97%
Thermo Scientific Chemicals
Malonaldehyde bis(diethyl acetal), 97%
CAS: 122-31-6 | C11H24O4 | 220.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA156735000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMalonaldehyde bis(diethyl acetal)
CAS122-31-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.