4-Iodoaniline, 99%
4-Iodoaniline, 99%
4-Iodoaniline, 99%
Thermo Scientific Chemicals
4-Iodoaniline, 99%
CAS: 540-37-4 | C6H6IN | 219.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA156610250
Price (MYR)
503.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
503.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Iodoaniline
CAS540-37-4
Health Hazard 1GHS Signal Word: Warning
Melting Point59.0°C to 64.0°C
ColorBeige to Brown-Gray