Phenethylamine, 99%
Phenethylamine, 99%
Phenethylamine, 99%
Thermo Scientific Chemicals
Phenethylamine, 99%
CAS: 64-04-0 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA156492500
Price (MYR)
413.00
Bottle
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
413.00
Bottle
Specifications
Chemical Name or MaterialPhenethylamine
CAS64-04-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre