Tribenzylamine, 99+%
Tribenzylamine, 99+%
Tribenzylamine, 99+%
Tribenzylamine, 99+%
Thermo Scientific Chemicals
Tribenzylamine, 99+%
CAS: 620-40-6 | C21H21N | 287.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Catalog number FSA156472500
Price (MYR)
695.00
BOT
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
695.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTribenzylamine
Name Note99+%
CAS620-40-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: