4-Hydroxy-3-methoxycinnamic acid, 99%
4-Hydroxy-3-methoxycinnamic acid, 99%
4-Hydroxy-3-methoxycinnamic acid, 99%
4-Hydroxy-3-methoxycinnamic acid, 99%
Thermo Scientific Chemicals
4-Hydroxy-3-methoxycinnamic acid, 99%
CAS: 1135-24-6 | C10H10O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA156361000
Price (MYR)
1,351.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,351.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Hydroxy-3-methoxycinnamic acid
Name Note99%
CAS1135-24-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning