2-Hydroxyethyl methacrylate, 97%, stabilized
2-Hydroxyethyl methacrylate, 97%, stabilized
2-Hydroxyethyl methacrylate, 97%, stabilized
2-Hydroxyethyl methacrylate, 97%, stabilized
Thermo Scientific Chemicals
2-Hydroxyethyl methacrylate, 97%, stabilized
CAS: 868-77-9 | C6H10O3 | 130.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number FSA156335000
Price (MYR)
409.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
409.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Hydroxyethyl methacrylate
Acidity2mg KOH/g max.
CAS868-77-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning