n-Heptadecane, 99%
n-Heptadecane, 99%
n-Heptadecane, 99%
Thermo Scientific Chemicals
n-Heptadecane, 99%
CAS: 629-78-7 | C17H36 | 240.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA156245000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Heptadecane
CAS629-78-7
Health Hazard 1GHS Signal Word: Danger
Melting Point22.0°C to 24.0°C