Glyoxal, pure, 40 wt.% solution in water
Glyoxal, pure, 40 wt.% solution in water
Glyoxal, pure, 40 wt.% solution in water
Glyoxal, pure, 40 wt.% solution in water
Thermo Scientific Chemicals
Glyoxal, pure, 40 wt.% solution in water
CAS: 107-22-2 | C2H2O2 | 58.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA156225000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS107-22-2, 7732-18-5
Specifications
Chemical Name or MaterialGlyoxal
Name Note40 wt.% Solution in Water
CAS107-22-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Suspected of causing genetic defects if inhaled.