Fluorene, 98+%
Fluorene, 98+%
Fluorene, 98+%
Thermo Scientific Chemicals
Fluorene, 98+%
CAS: 86-73-7 | C13H10 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA156135000
Price (MYR)
1,419.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,419.00
BOT
Add to cart
Specifications
Chemical Name or MaterialFluorene
Name Note98%
CAS86-73-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Very toxic to aquatic life with long lasting effects.