3-Ethoxy-4-hydroxybenzaldehyde, 97%
3-Ethoxy-4-hydroxybenzaldehyde, 97%
3-Ethoxy-4-hydroxybenzaldehyde, 97%
3-Ethoxy-4-hydroxybenzaldehyde, 97%
Thermo Scientific Chemicals
3-Ethoxy-4-hydroxybenzaldehyde, 97%
CAS: 121-32-4 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA156030050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Ethoxy-4-hydroxybenzaldehyde
Name Note97%
CAS121-32-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning