2-Ethoxyethanol, 99%, extra pure
2-Ethoxyethanol, 99%, extra pure
2-Ethoxyethanol, 99%, extra pure
2-Ethoxyethanol, 99%, extra pure
Thermo Scientific Chemicals
2-Ethoxyethanol, 99%, extra pure
CAS: 110-80-5 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA156020025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Ethoxyethanol
CAS110-80-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0