1,5-Dibromopentane, 97%
1,5-Dibromopentane, 97%
1,5-Dibromopentane, 97%
1,5-Dibromopentane, 97%
Thermo Scientific Chemicals
1,5-Dibromopentane, 97%
CAS: 111-24-0 | C5H10Br2 | 229.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA155992500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 5-Dibromopentane
Name Note97%
CAS111-24-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning