Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA155895000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethylamine hydrochloride
Name Note99%
CAS660-68-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: