3-Nitrobenzoyl chloride, 98%
3-Nitrobenzoyl chloride, 98%
3-Nitrobenzoyl chloride, 98%
3-Nitrobenzoyl chloride, 98%
Thermo Scientific Chemicals
3-Nitrobenzoyl chloride, 98%
CAS: 121-90-4 | C7H4ClNO3 | 185.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA155840050
Price (MYR)
202.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
202.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Nitrobenzoyl chloride
Name Note98%
CAS121-90-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger