Alizarin Complexone dihydrate, pure, indicator grade
Alizarin Complexone dihydrate, pure, indicator grade
Alizarin Complexone dihydrate, pure, indicator grade
Alizarin Complexone dihydrate, pure, indicator grade
Thermo Scientific Chemicals
Alizarin Complexone dihydrate, pure, indicator grade
CAS: 455303-00-1 | C19H19NO10 | 421.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA155830050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Absorbance0.42 min. (lambda max, after 1 hr.) (2mL of a 0.2% soln. in 0.02M NaOH + 5mL acetate buffer (pH 5.1) diluted with H2O to 100mL), (1cm cell vs. water)
Chemical Name or MaterialAlizarin Complexone dihydrate
Melting Point180.0°C to 185.0°C
CAS455303-00-1
Health Hazard 1Warning
View more
Indicator
RUO – Research Use Only