3-Pentanone, 98%, pure
3-Pentanone, 98%, pure
3-Pentanone, 98%, pure
3-Pentanone, 98%, pure
Thermo Scientific Chemicals
3-Pentanone, 98%, pure
CAS: 96-22-0 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA155815000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Pentanone
Name Note98%
CAS96-22-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger