n-Heptylamine, 99+%
n-Heptylamine, 99+%
n-Heptylamine, 99+%
Thermo Scientific Chemicals
n-Heptylamine, 99+%
CAS: 111-68-2 | C7H17N | 115.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA155461000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Heptylamine
Name Note99+%
CAS111-68-2
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger