1-Dodecanol, 98%
1-Dodecanol, 98%
1-Dodecanol, 98%
Thermo Scientific Chemicals
1-Dodecanol, 98%
CAS: 112-53-8 | C12H26O | 186.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
1 L
Catalog number FSA155455000
Price (MYR)
279.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
279.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Dodecanol
CAS112-53-8
CAS Min %97.5
Health Hazard 1Warning
CAS Max %100.0