alpha,alpha'-Dibromo-m-xylene, 97%
alpha,alpha'-Dibromo-m-xylene, 97%
alpha,alpha'-Dibromo-m-xylene, 97%
alpha,alpha'-Dibromo-m-xylene, 97%
Thermo Scientific Chemicals
alpha,alpha'-Dibromo-m-xylene, 97%
CAS: 626-15-3 | C8H8Br2 | 263.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA155050250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialα, α'-Dibromo-m-xylene
CAS626-15-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0