Cyclopropylamine, 99%
Cyclopropylamine, 99%
Cyclopropylamine, 99%
Cyclopropylamine, 99%
Thermo Scientific Chemicals
Cyclopropylamine, 99%
CAS: 765-30-0 | C3H7N | 57.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
Packaging:
Glass bottle
Catalog number FSA154880500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCyclopropylamine
CAS765-30-0
Health Hazard 1GHS Signal Word: Danger
Melting Point-50.0°C