1,4-Cyclohexadiene, 97%, stabilized
1,4-Cyclohexadiene, 97%, stabilized
1,4-Cyclohexadiene, 97%, stabilized
1,4-Cyclohexadiene, 97%, stabilized
Thermo Scientific Chemicals
1,4-Cyclohexadiene, 97%, stabilized
CAS: 628-41-1 | C6H8 | 80.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA154821000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 4-Cyclohexadiene
Name NoteStabilized
CAS628-41-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger